Chrysoidine G for microscopy, Sigma-Aldrich®
:532-82-1
:
:C₁₂H₁₃ClN₄
:MFCD00012976
:248.714 g/mol
:235 °C (dec., lit.)
:M-Clarity™ quality level = MQ100
:208-545-8
:GHS08
:GHS05
:GHS07
:GHS09
:Room Temperature
- Appearance (colour):Red to very dark red and purple to very dark purple and brown to very dark brown and very dark red-brown
- Appearance (form):Powder or crystals
- Thin layer chromatogr.:Corresponds to standard
- Infrared spectrum:Conforms to structure
- Wavelength (1) (UV):444.0 - 454.0 nm
- Solvent (UV):Water