Order Entry
Finland
ContactUsLinkComponent
Basacryl Red GL 19% (dye content), Sigma-Aldrich®
Basacryl Red GL 19% (dye content), Sigma-Aldrich®
Catalog # 306525-25G
CAS Number:  
undefined
Basacryl Red GL 19% (dye content), Sigma-Aldrich®
Catalog # 306525-25G
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :368.88 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:42373-04-6
Formula:C₁₉H₁₇ClN₄S
MDL Number:MFCD00013337