Order Entry
Finland
ContactUsLinkComponent
Papanicolaou's solution EA 65, Sigma-Aldrich®
Papanicolaou's solution EA 65, Sigma-Aldrich®
Catalog # HT40432-1L
CAS Number:  
Papanicolaou's solution EA 65, Sigma-Aldrich®
Catalog # HT40432-1L
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
ADR:3,II
 :GHS08
 :GHS07
Storage Temperature:Ambient
 :1