Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Keratin azure
Keratin azure, Sigma-Aldrich®
Keratin azure, Sigma-Aldrich®
Keratin azure, Sigma-Aldrich®
Catalog # K8500-5G
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Expand image
Keratin azure, Sigma-Aldrich®
Catalog # K8500-5G
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
Pk:
5 g
Seq:
3173
Specifications
About this item
Product Details
:
Storage Temperature:
Ambient
MDL Number:
MFCD00131435
Product Details