Order Entry
Finland
ContactUsLinkComponent
Thymol iodide, Sigma-Aldrich®
Thymol iodide, Sigma-Aldrich®
Catalog # T2763-25G
CAS Number:  
undefined
Thymol iodide, Sigma-Aldrich®
Catalog # T2763-25G
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :550.219 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:552-22-7
Formula:C₂₀H₂₄I₂O₂
MDL Number:MFCD00026409