PDAM, Sigma-Aldrich®
Catalog # P6863-25MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:242.27 g/mol
Storage Temperature:Freezer
CAS Number:78377-23-8
Formula:C₁₇HN₂
MDL Number:MFCD00467268