Order Entry
Finland
ContactUsLinkComponent
Ponceau S solution, Sigma-Aldrich®
Ponceau S solution, Sigma-Aldrich®
Catalog # P7170-1L
CAS Number:  
Ponceau S solution, Sigma-Aldrich®
Catalog # P7170-1L
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :760.56 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:6226-79-5
Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:MFCD00003892