Pararosaniline, crystalline, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:467-62-9
:25620-78-4
Formula:C₁₉H₁₉N₃O
MDL Number:MFCD00036222
:305.38 g/mol
Melting Pt:205 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
:GHS08
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Light Red to Very Dark Purple
- Appearance (Form):Powder
- Solubility (Colour):Lt. Pink to Red to Very Dark Purple
- Solubility (Turbidity)
1 mg/ml, EtOH:Clear to Hazy - Infrared spectrum:Conforms
- Carbon:70.2 - 79.2 %
- Nitrogen:12.9 - 14.6 %