Ponceau Xylidine ≥60 % (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:3761-53-3
:
Formula:C₁₈H₁₆N₂O₇S₂
MDL Number:MFCD00003897
:480.42 g/mol
:M-Clarity™ quality level = MQ200
:GHS08
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Red
- Appearance (Form):Powder
- Solubility (Colour):Red
- Solubility (Turbidity)
10 mg/mL, H2O:Clear - Nitrogen:3.5 - 5.8 %
- Dye Content Calculated:≥ 60
- UV (Lambda max):503 - 508 nm
- UV (EmM at Lambda max)
in H2O:18 - 22