Rose bengal diacetate, Sigma-Aldrich®
Catalog # R8401-5MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:1057.75 g/mol
Storage Temperature:Freezer
CAS Number:61738-01-0
Formula:C₂₄H₈Cl₄I₄O₇