Ruthenium(III) chloride oxide, Technical Grade, Sigma-Aldrich®
:11103-72-3
:
:H₄₂Cl₆N₁₄O₂Ru₃
:MFCD00011479
:786.354 g/mol
:M-Clarity™ quality level = MQ200
:GHS07
:Room Temperature
- Appearance (Form):Powder
- Solubility (Colour):10 mg/ml water
- Solubility (Method):10 mg/ml water
- Wavelength (1) (Uv):533 - 534 nm
- Extinction (1%/1cm) (1) (Uv):Jun-03
- Solvent (UV):Ammonium Acetate 0.1 M
- Assay (Icp):34 - 39 % RUTHENIUM