N-Carboxymethyl-6-(2,2-dicyanovinyl)-1,2,3,4-tetrahydroquinoline ≥95% (by HPLC), Sigma-Aldrich®
Catalog # C6206-25MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:267.28 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:47072-52-6
Formula:C₁₅H₁₃N₃O₂
MDL Number:MFCD00274409