Aniline blue solution 2.5% in 2% acetic acid for dye, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:28631-66-5
:8004-91-9
Formula:C₃₂H₂₅N₃Na₂O₉S₃
MDL Number:MFCD00036130
:737.74 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
Specification Test Results
- Appearance:Dark Blue, Sediment Free
- UV/VIS Absorbance:0.21 - 0.37
- Lambda max:597-603 nm