Eosin Y (yellowish) alcoholic solution, Sigma-Aldrich®
Catalog # HT110116-500ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
ADR:3,II
:691.86 g/mol
:GHS02
:GHS05
Storage Temperature:Ambient
:1
CAS Number:17372-87-1
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040