Order Entry
Finland
ContactUsLinkComponent
Haematoxylin Gill No. 3, Sigma-Aldrich®
Haematoxylin Gill No. 3, Sigma-Aldrich®
Catalog # GHS332-1L
CAS Number:  
Haematoxylin Gill No. 3, Sigma-Aldrich®
Catalog # GHS332-1L
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :302.28 g/mol
 :GHS08
 :GHS05
Storage Temperature:Ambient
 :1
CAS Number:517-28-2
 :208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111