Fast blue BB salt ≥80% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:5486-84-0
:
Formula:C₃₄H₃₆Cl₄N₆O₆Zn
MDL Number:MFCD00074765
Melting Pt:157 °C
:M-Clarity™ quality level = MQ200
:831.89 g/mol
EINECS:226-817-4
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Solubility (Turbidity), c = 10 mg/ml; Water:Clear to Very Hazy
- Solubility (Colour):Yellow to Very Dark Yellow and Brown and Yellow-Brown
- Nitrogen:≥8.0%
- Dye Content Calculated, Based on Nitrogen Content:≥80%