PAS (Periodic Acid Schiff) staining kit, Sigma-Aldrich®
Catalog # 395B-1KT
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
Specifications
About this item
:
ADR:9,II
:GHS05
:GHS07
Storage Temperature:Refrigerator
:1