Order Entry
Finland
ContactUsLinkComponent
Fetal Hemoglobin Kit, Sigma-Aldrich®
Fetal Hemoglobin Kit, Sigma-Aldrich®
Catalog # 285D-1KT
CAS Number:  
Fetal Hemoglobin Kit, Sigma-Aldrich®
Catalog # 285D-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ500 (more info.)
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
 :1