Anti-Coronavirus (SARS-CoV-2) spike S1 Mouse Monoclonal Antibody
Supplier: MP Biomedicals
Certificates
About this item
Specifications
- Catalog No:
- 08720301.
- 08720302.
- Antigen name:
- Coronavirus (SARS-CoV-2) spike S1
- Coronavirus (SARS-CoV-2) spike S1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1