
Tiamulin fumarate
Catalog # ACRO462820010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:143 to 152 °C
- Loss on drying:≤0.5 %
- HPLC:≥97.5 %
- Titration with NaOH:18.5 to 19.5 % (Fumaric acid)
Specification Test Results
About this item
:
Boiling Pt:563 °C (760 mmHg)
Melting Pt:147…148 °C
Flash Pt:>110 °C
:609.819 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:55297-96-6
Formula:C₃₂H₅₁NO₈S
MDL Number:MFCD00145407