Order Entry
Finland
ContactUsLinkComponent
Copper test kit for use with spectrophotometer, Vacu-vials®
Copper test kit for use with spectrophotometer, Vacu-vials®
Catalog # CHETK-3503
Supplier:  CHEMetrics
CAS Number:  
Copper test kit for use with spectrophotometer, Vacu-vials®
Catalog # CHETK-3503
Supplier:  CHEMetrics
CAS Number:  
Specifications
  • Chemical method:
    Bathocuproine
  • Pk:
    1 KIT
  • Description:
    Copper
  • Tests per kit:
    30
  • Range:
    0 - 12,00 ppm
Specifications
About this item
The method measures total soluble copper as ppm (mg/L) Cu. The test kits are applicable for analysis of drinking water, surface waters, groundwater, wastewater and seawater.
  • Premixed
  • Premeasured
  • Precise
Ordering information: This kit requires the use of a CHEMetrics Direct-Readout Photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, 25 mL sample cup, ampoule blank, and instructions.