Wipes, KIMTECH PREP® SCOTTPURE™
Catalog # KIMB61930
Supplier: KIMBERLY CLARK
Specifications
- Packed:Center-Pull Rolls (225 sheets per roll)
- Colour:White
- Description:SCOTTPURE™ wipes
- Pk:6 Roll
Specifications
About this item
KIMTECH PREP® SCOTTPURE™ Wipers are designed for wiping tasks where precision and reliability are essential.
Low-lint rayon/polyester spunlace material for cloth-like softness. Engineered for excellent absorbency and low-lint wiping. Solvent resistant. No binders or surfactants. Applications include aerospace, clean manufacturing areas, DVD manufacturing, and printing.
Ordering information: Supplied without dispenser.