Order Entry
Finland
ContactUsLinkComponent
Cleaning cloths, undyed, WYPALL*
Cleaning cloths, undyed, WYPALL*
Catalog # KIMB38715
Supplier:  KIMBERLY CLARK
undefined
Cleaning cloths, undyed, WYPALL*
Catalog # KIMB38715
Supplier:  KIMBERLY CLARK
Specifications
  • Packed:
    20/pack, 1 pack/case
  • Colour:
    White
  • L×W:
    400×400 mm
  • Pk:
    25
Specifications
About this item
These microfibre cloths lift and trap surface dirt. Ideal for cleaning in offices, healthcare, and hospitality.
  • Perfect for use in transportation manufacturing
  • Suitable for a wide range of cleaning tasks
  • Washable
These microfibre cloths are designed with a high quality polyester/polyamide that doesn't fray making them perfect for automotive valet tasks.