Safety canisters
About this item
These canisters are made of high-quality PE-HD material with high chemical resistance for use in the laboratory for safe disposal of chromatographic solvents or other liquid laboratory waste for the highest level of work safety in the laboratory.
- Made of very robust HDPE, which is very resistant to almost all solvents commonly used in the laboratory
- Smallest canisters have a volume of only 5 L and are ideally suited for use on tables or smaller storage areas
- Larger canisters are available with volumes of 10, 12, 20, 30 up to 60 L, which can be deposited under tables or in safety cabinets
- Thread types S55, S60/61, S65, S70/71, S90 and S95 are commonly used in the laboratory, they are all compatible with the SCAT SafetyWasteCaps range for the safe disposal of volatile hazardous substances
- Special versions available with floats for level indication or with internal fluorination for double protection of the contents, both for permeation from outside to inside and for diffusion from inside to outside
The wide range of non-electrically conductive canisters includes a variety of designs for individual use in the laboratory for the disposal of solvents in combination with the SCAT SafetyWasteCaps to increase your laboratory safety.
Most SCAT canisters are UN-Y or UN-X approved, which ensures safe storage or safe transport of eluents.
18 Options Available Below
Product Details & Documents
These canisters are made of high-quality PE-HD material with high chemical resistance for use in the laboratory for safe disposal of chromatographic solvents or other liquid laboratory waste for the highest level of work safety in the laboratory.
- Made of very robust HDPE, which is very resistant to almost all solvents commonly used in the laboratory
- Smallest canisters have a volume of only 5 L and are ideally suited for use on tables or smaller storage areas
- Larger canisters are available with volumes of 10, 12, 20, 30 up to 60 L, which can be deposited under tables or in safety cabinets
- Thread types S55, S60/61, S65, S70/71, S90 and S95 are commonly used in the laboratory, they are all compatible with the SCAT SafetyWasteCaps range for the safe disposal of volatile hazardous substances
- Special versions available with floats for level indication or with internal fluorination for double protection of the contents, both for permeation from outside to inside and for diffusion from inside to outside
The wide range of non-electrically conductive canisters includes a variety of designs for individual use in the laboratory for the disposal of solvents in combination with the SCAT SafetyWasteCaps to increase your laboratory safety.
Most SCAT canisters are UN-Y or UN-X approved, which ensures safe storage or safe transport of eluents.
Product Family Options
UN-certified
Product Information
- CapacityColourThreadTypeW×D×HAvailabilityPrice
- 590-0647Safety canisterSafety canister2,5 LNaturalGL 45UN-Y approval115×150×210 mmLog in for availability1 * 1 Items22,10 €
- 590-0648Safety canisterSafety canister5 LNaturalGL 45UN-Y approval150×195×250 mmLog in for availability1 * 1 Items36,50 €
- 590-0655Safety canister
- SCTV107933Safety canisterSafety canister5 LNaturalS 55UN-X approval; with inner fluorination145×190×250 mmLog in for availability1 * 1 Items30,40 €
- 590-0654Safety canister
- SCTV108175Safety canister
- SCTV108227Safety canisterSafety canister5 LNaturalS 60/61UN-Y approval164×185×235 mmLog in for availability1 * 1 Items31,60 €
- 590-0574Safety canister
- 590-0649Safety canisterSafety canister10 LNaturalGL 45UN-Y approval190×230×305 mmLog in for availability1 * 1 Items40,10 €
- 590-0652Safety canisterSafety canister10 LNaturalS 55UN-Y approval185×225×305 mmLog in for availability1 * 1 Items31,60 €
- SCTV108189Safety canisterSafety canister10 LNaturalS 55UN-Y approval, with inner fluorination190×230×310 mmLog in for availability1 * 1 Items60,80 €
- 590-0710Safety canisterSafety canister10 LNaturalS 90UN-Y approval195×195×380 mmLog in for availability1 * 1 Items36,50 €
- 554-1849Safety canisterSafety canister12 LNaturalS 60/61UN-Y approval200×235×350 mmLog in for availability1 * 1 Items26,70 €
- 590-0653Safety canisterSafety canister20 LNaturalS 60/61UN-Y approval260×289×390 mmLog in for availability1 * 1 Items36,80 €
- 590-0656Safety canisterSafety canister30 LNaturalS 60/61UN-Y approval290×380×400 mmLog in for availability1 * 1 Items36,50 €
- SCTV107710Safety canisterSafety canister60 LNaturalS 70/71UN-Y approval330×370×635 mmLog in for availability1 * 1 Items97,40 €
Non UN-certified
Product Information
- CapacityColourThreadTypeW×D×HAvailabilityPrice
- 590-0741Safety canisterSafety canister20 LNaturalS 60/61With floater260×285×455 mmLog in for availability1 * 1 Items133,00 €
- SCTV108115Safety canisterSafety canister30 LNaturalS 60/61 and GL 452 openings290×380×400 mmLog in for availability1 * 1 Items170,00 €
Replacement lid
Product Information
- Catalog #ColourThreadDescriptionAvailabilityPrice