About this item
TitroLine® 7000 automatic titrators are used in the laboratories of water supply companies, wastewater treatment plants, and the food and chemical industries. Typical applications include analysis of alkalinity and carbon dioxide, nitrogen (Kjeldahl), chloride, COD, acid number, iodine number and others. The Titronic® 500 piston burette is used in all industry sectors. Applications include all manual titration, dosing tasks and sample preparation.
- Brilliant, high contrast, TFT display
- Compact exchangeable units (5/10/20/50 ml) can store all reagent-specific data plus the unit size (TitroLine® 7000 only)
- Connection of stirrer, keyboard, printer, balance, PC and USB storage media can be done via three USBs and two RS232 ports
- Standard methods and formulae allow easy generation of up to 50 user-defined methods
- Common functions: Manual titration, dosing tasks and formulae for miscellaneous calculations
- Thanks to their flexibility (exchangeable units), they are perfect for customers who require a titrator for many different applications
TitroLine® 6000 basic unit: Supplied with magnetic stirrer TM 235, exchangeable unit (10, 20 or 50 ml), incl. brown glass bottle for titration reagent, GL 45 and S 40 bottle adapters, hoses, drip off tube and titration tip.
TitroLine® 7000 basic unit: Supplied with magnetic stirrer TM 235, exchangeable unit (10, 20 or 50 ml), incl. brown glass bottle for titration reagent, GL 45 and S 40 bottle adapters, hoses, drip off tube and titration tip.
TitroLine® 7000 basic unit: Supplied with TM 235 magnetic stirrer, exchangeable unit (10, 20 or 50 ml), incl. brown glass bottle for titration reagent, GL 45 and S 40 bottle adapters, hoses, drip off tube and titration tip.
Specifications
- Catalog No:
- 661-0114
- 661-0115
- 661-0116
- 661-0117
- SCOI285220170
- 661-0121
- Model:
- TitroLine® 7000
- TitroLine® 7000
- TitroLine® 7000
- Titronic® 500
- TitroLine® 7000
- Titronic® 500
- Display:
- High contrast TFT
- High contrast TFT
- High contrast TFT
- High contrast TFT
- High contrast TFT
- High contrast TFT
- Dosing volume:
- 10 / 20 / 50 ml
- 10 / 20 / 50 ml
- 10 / 20 / 50 ml
- —
- 10 / 20 / 50 ml
- 20 ml
- Method storage:
- 50
- 50
- 50
- 5
- 50
- 5
- Functions:
- pH/mV measuring input with reference input;
titration to mV and pH endpoint;
dynamic and linear titration to equivalence point (EQ), mV and pH;
dead-stop titration;
pH-stat titration;
ideal for non aqueous titration - pH/mV measuring input with reference input;
titration to mV and pH endpoint;
dynamic and linear titration to equivalence point (EQ), mV and pH;
dead-stop titration;
pH-stat titration;
ideal for non aqueous titration - pH/mV measuring input with reference input;
titration to mV and pH endpoint;
dynamic and linear titration to equivalence point (EQ), mV and pH;
dead-stop titration;
pH-stat titration;
ideal for non aqueous titration - Preparation of solvents (manual or automatic with connected balance)
- pH/mV measuring input with reference input;
titration to mV and pH endpoint;
dynamic and linear titration to equivalence point (EQ), mV and pH;
dead-stop titration;
pH-stat titration;
ideal for non aqueous titration - Preparation of solvents (manual or automatic with connected balance)
- Measuring input electrode:
- 2×4 mm socket
- 2×4 mm socket
- 2×4 mm socket
- —
- 2×4 mm socket
- —
- Connections:
- 3×USB, 2×RS232
- 3×USB, 2×RS232
- 3×USB, 2×RS232
- 3×USB, 2×RS232
- 3×USB, 2×RS232
- 3×USB, 2×RS232
- W×D×H:
- 153×296×450 mm
- 153×296×450 mm
- 153×296×450 mm
- 153×296×450 mm
- 153×296×450 mm
- 153×296×450 mm
- Weight:
- 3,5 kg
- 3,5 kg
- 3,5 kg
- 3,5 kg
- 3,5 kg
- 3,5 kg