Specifications
- Antibody type:Primary
- Antigen name:BDNF/NT-3 growth factors receptor
- Antigen symbol:NTRK2
- Clonality:Monoclonal
- Clone:BS379
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Epitope:Extracellular, amino acids 179-190 of human TrkB
- Cross adsorption:No
- Form:Lyophilized
- Gene ID:Q16620 (NTRK2_HUMAN)
- Antigen synonyms:GP145-TrkB|Tropomyosin-related kinase B|TrkB tyrosine kinase|Neurotrophic tyrosine kinase receptor type 2
- Amino acid number:179 to 190
- Storage temperature:Store lyophilized antibody at 2-8C. After reconstitution divide into aliquots and store at -20C for long-term storage. Store at 2-8C short-term (up to 4 weeks). Avoid repetitive freeze/thaw cycles.
- Shipping temperature:RT
- Immunogen:Synthetic peptide immunogen, ESSKNIPLANLQ
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The protein named TrkB (also named Neurotrophic tyrosine kinase receptor type 2 (NTRK2), GP145-TrkB or Tropomyosin-related kinase B is a receptor tyrosine kinase involved in the development and the maturation of the central and the peripheral nervous systems and is important in the regulation of neuron survival, proliferation, migration, differentiation, and synapse formation and plasticity. TrkB may also play a role in neutrophin-dependent calcium signaling in glial cells and mediate communication between neurons and glia. TrkB is the primary receptor for BDNF (brain-derived neurotrophic factor. TrkB also binds NT4 and NT3 but less efficiently. (Reference: www.uniprot.org).
Applications: Flow Cytometry (5-10 ug/mL): Tested on human and rodent cell lines. Cell staining can be performed under native conditions on ice, or on fixed cells with up to 4% formaldehyde. Other applications have not been tested. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application notes: Flow Cytometry (5-10 ug/mL): Tested on human and rodent cell lines. Cell staining can be performed under native conditions on ice, or on fixed cells with up to 4% formaldehyde.
Other applications have not been tested. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.