Order Entry
Finland
ContactUsLinkComponent
Bacteria genomicPrep mini spin kit, illustra™
Bacteria genomicPrep mini spin kit, illustra™
Supplier:  Cytiva
Certificates
undefined
Bacteria genomicPrep mini spin kit, illustra™
Supplier:  Cytiva

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Bacterial DNA
  • Sample type:
    Bacteria
  • Target molecule:
    DNA
  • Description:
    illustra™ bacteria genomicPrep Mini Spin Kit

Specifications