Hygiene mask, fine twill cotton
Catalog # 113-9934
Supplier: RENTEX
Specifications
- Type:with elastic bands
- Colour:White
- Size:180x100 mm
- Pk:250
Specifications
About this item
Made from 100% one layer cotton, this fine twill mask can be washed on higher temperatures for repeated use.
- Single layer, 100% cotton construction
- Comfortable fit with elastic bands around ears
- Washable on higher temperatures
- Non sterile
Designed to provide additional mouth/nose protection (AMNP), these cotton masks should serve only as an alternative solution for protection. These masks may help to reduce infection, however, they are neither tested nor certified, and are not sterile.
Packaging: Masks are packed 50 per plastic bag, 12 bags per case.