Order Entry
Finland
ContactUsLinkComponent
Welding helmet, BizWeld Plus, PW65
Welding helmet, BizWeld Plus, PW65
Catalog # PRTWPW65BKR
Supplier:  Portwest
Welding helmet, BizWeld Plus, PW65
Catalog # PRTWPW65BKR
Supplier:  Portwest
Specifications
  • Description:
    Welding helmet, black
  • Material:
    PP/PE
  • Model:
    PW65
  • Pk:
    1
Specifications
About this item
Auto-darkening welding helmet, which switches from a light to dark state when welding activity starts.
  • Wheel ratchet size adjustment for easy fitting
  • Solar cell power with two built-in lithium back-up batteries and a working time of over 5000 hours
  • Auto-darkening, switches from light to dark when the welding process starts
  • Suitable for a wide range of applications
  • Suitable to be worn with disposable respirators
  • Retail box which aids presentation for retail sales
Response time from light to dark state is 0,04 milliseconds; return from dark to clear state in 0,03 to 0,04 second.
Power is supplied by solar cells with 2 built-in lithium batteries to offer power back-up for longer service life. No need for replacement batteries.
Standard: EN 166 1 S, EN 175 F, EN 379 4 / 9-13 ; 1/1/1/2.
Replacement Lens
Product Information