Order Entry
Finland
ContactUsLinkComponent
Endurance visor replacement, PW56
Endurance visor replacement, PW56
Catalog # PRTWPW56CLR
Supplier:  Portwest
Endurance visor replacement, PW56
Catalog # PRTWPW56CLR
Supplier:  Portwest
Specifications
  • Description:
    Replacement visor, clear
  • Material:
    Polycarbonate
  • Model:
    PW56
  • Pk:
    1
Specifications
About this item
Impact resistant polycarbonate replacement visor is suitable for endurance helmets.
  • Anti-scratch coating for added durability
  • Anti-fog coating for additional comfort to the wearer
Suitable for PW54, PW55 and PS55 helmets.
Certifications: Standard: EN 166 1 FT.
Packaging: Individually packed for vending machines.