General purpose gloves, Dermi Pro Glove, AP30
: Portwest
These fully dipped nitrile coated gloves with foam nitrile coating in the palm area provides comfort and touch sensitivity. An elastic knit cuff holds it firmly on the arm for enhanced fit. The nitrile coating offers excellent liquid barrier while the superior foam dip ensures excellent grip even in oily environments.
- Super abrasion and tear resistance
- Fully coated for maximum liquid protection
- Nitrile foam coating for excellent grip in wet and dry conditions
- Ergonomic design to reduce hand fatigue
- 13 gauge liner for a perfect fit
- Retail tag which aids presentation for retail sales
The general handling and grip glove category is still one of the most popular collections. A carefully selected range of coatings, including latex, nitrile, polyurethane and PVC, provides a broad range of styles suited to a diverse range of tasks.
: AS/NZS 2161 .2; AS/NZS 2161 .3; ANSI/ISEA 105 - 2016 CUT Level (A1); EN 420 (Dexterity 5); EN388:2016 +A1:2018 - (4.1.2.1.X).
- Catalog No:
- PRTWAP30O8RXXL
- PRTWAP30O8RXL
- PRTWAP30O8RS
- PRTWAP30O8RM
- PRTWAP30O8RL
- Model:
- AP30
- AP30
- AP30
- AP30
- AP30
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Material:
- Nitrile/nylon
- Nitrile/nylon
- Nitrile/nylon
- Nitrile/nylon
- Nitrile/nylon
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- EN norm:
- EN 420 (Dexterity 5); EN388:2016 +A1:2018 - (4.1.2.1.X)
- EN 420 (Dexterity 5); EN388:2016 +A1:2018 - (4.1.2.1.X)
- EN 420 (Dexterity 5); EN388:2016 +A1:2018 - (4.1.2.1.X)
- EN 420 (Dexterity 5); EN388:2016 +A1:2018 - (4.1.2.1.X)
- EN 420 (Dexterity 5); EN388:2016 +A1:2018 - (4.1.2.1.X)