Order Entry
Finland
ContactUsLinkComponent
 
Analytical standard, Sarafloxacin-hydrochloride hydrate, reference material, Fluka™
  HONCR1715-100MG
 :  
Analytical standard, Sarafloxacin-hydrochloride hydrate, reference material, Fluka™
  HONCR1715-100MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    91296-87-6
  • Description:
    Sarafloxacin-hydrochloride hydrate
  • Description:
    Sarafloxacin-hydrochloride hydrate
  • Form:
    Neat