Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Analytical standard, Linuron, reference material, Fluka™
Analytical standard, Linuron, reference material, Fluka™
Analytical standard, Linuron, reference material, Fluka™
HONCR1793-100MG
:
Honeywell Chemicals
:
Analytical standard, Linuron, reference material, Fluka™
HONCR1793-100MG
:
Honeywell Chemicals
:
Pack type:
Glass bottle
Pk:
100 mg
Standards:
Reference material
CAS No.:
330-55-2
Description:
Linuron
Description:
Linuron
Form:
Neat