Ribbons (heavy-duty), for BBP3X/S3XXX/i3300 printers
Supplier: Brady
Certificates
About this item
Drop and Click print ribbons for quick and easy supply changes.
- Resin ribbon formulation
Brady series R6600 thermal transfer ribbon is a resin formulation which offers excellent solvent and friction resistance when used with Brady heat-shrinkable tubing materials, including B-342, B-7641, B-7642 and B-7646.
Certifications: RoHS compliant with Directive 2011/65/EU and 2015/863. Part does not contain an SVHC.