Order Entry
Finland
ContactUsLinkComponent
Industrial label printers, BradyPrinter i7100
Industrial label printers, BradyPrinter i7100
Supplier:  Brady
Certificates
undefined
Industrial label printers, BradyPrinter i7100
Supplier:  Brady
Specifications
  • Catalog No:
  • BRDY149047
  • BRDY198613
  • BRDY149046
  • Memory:
  • 256 MB RAM, 50 MB IFFS Flash, SD card,
    slot up to 512 GB, internal
  • 256 MB RAM, 50 MB IFFS Flash, SD card,
    slot up to 512 GB, internal
  • 256 MB RAM, 50 MB IFFS Flash, SD card,
    slot up to 512 GB, internal
  • Resolution:
  • 300 or 600 dpi
  • 300 or 600 dpi
  • 300 or 600 dpi
  • Dimensions:
  • 252W×288H×460D mm
  • 252W×288H×460D mm
  • 252W×288H×460D mm
  • Weight:
  • 10 kg
  • 10 kg
  • 10 kg