Order Entry
Finland
Orders LinkContactUsLinkComponent
2'-Deoxyguanosine monohydrate 98+% (dry basis)

2'-Deoxyguanosine monohydrate 98+% (dry basis)

10 g
Catalog # J14304.09
2'-Deoxyguanosine monohydrate 98+% (dry basis)
Catalog # J14304.09
CAS Number:  312693-72-4
2'-Deoxyguanosine monohydrate 98+% (dry basis)
2'-Deoxyguanosine monohydrate 98+% (dry basis)

10 g

CAS Number:  312693-72-4
Price
1 350,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one hydrate
  • CAS Number:312693-72-4
  • Formula:C₁₀H₁₃N₅O₄·H₂O
  • MDL Number:MFCD00150760
  • Molecular Weight:285.26 g/mol
  • Storage Temperature:Ambient
  • EINECS:213-505-8
Specifications

Specification Test Results

  • Form:
    White crystalline powder
  • Assay (unspecified):
    ≥ 98% (dry basis)
  • Assay (special): Absorbance Ratios: A250/A260, A280/A260:
    1.12 to 1.18, 0.65 to 0.69 (pH 7.0)
  • Assay (special): Extinction Coefficient:
    13.7 X 103 ± 4% (pH 7.0)
  • Identification (FTIR):
    Nujol mull matches reference scan
  • Water Content (Karl Fischer Titration):
    ≤ 7.0%
  • lambda-max (UV-Vis):
    253 ± 2 nm (pH 7.0)
  • Thin Layer Chromatography:
    Homogenous

Specifications

  • Pk:
    10 g

Specifications