Order Entry
Finland
ContactUsLinkComponent
2,4,6-Triphenyl-1,3,5-triazine 99%
Irritant
2,4,6-Triphenyl-1,3,5-triazine 99%
Certificates
undefined
Irritant
2,4,6-Triphenyl-1,3,5-triazine 99%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    231 to 236 °C
  • HPLC:
    ≥98.5 %

Specification Test Results

Related Information