

Titanium(IV) isopropoxide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:546-68-9
:
Formula:C₁₂H₂₈O₄Ti
MDL Number:MFCD00008871
Merck Index:14,9480
Boiling Pt:232 °C
Melting Pt:16…20 °C
Density:0.955
Flash Pt:46 °C (115 °F)
UN:2413
ADR:3,III
:284.23 g/mol
EINECS:208-909-6
:GHS02
:GHS07
Storage Temperature:Ambient