
Titanium carbide, nanopowder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12070-08-5
:
Formula:CTi
MDL Number:MFCD00011268
Boiling Pt:4820 °C
Melting Pt:∼ 3140 °C
Density:4.93
Storage Temperature:Ambient
EINECS:235-120-4
UN:3178
ADR:4.1,III
:59.89 g/mol
:GHS02