Strontium titanate S.A. 12.5-25 m²/g
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12060-59-2
:
Formula:O₃SrTi
MDL Number:MFCD00049554
:183.5 g/mol
Melting Pt:2060 °C
Density:4.810
Storage Temperature:Ambient
EINECS:235-044-1