


1,2-Epoxycyclohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:286-20-4
:
Formula:C₆H₁₀O
MDL Number:MFCD00005162
:98.14 g/mol
Boiling Pt:129 °C (1013 hPa)
Melting Pt:–27 °C
Density:0.97 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-007-7
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
UN:2920
:GHS02
:GHS06
:GHS05
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-27 °C
- Molar Mass:98.14 g/mol
- Boiling Point:131 - 132 °C (1013 hPa)
- Vapor Pressure:12 hPa (20 °C)
- Flash Point:24 °C
- Refractive Index:1.4515 (20 °C, 589 nm)
- Explosion Limit:1.2 - 6.5 %(V)
- Density:0.97 g/cm3 (20 °C)
- Ignition Point:335 °C