Order Entry
Finland
Orders LinkContactUsLinkComponent
 

4'-Cyclohexylacetophenone

Glass bottle for solids, 25 g
Catalog # 8.14012.0025
Supplier:   Merck
4'-Cyclohexylacetophenone
Catalog # 8.14012.0025
Supplier:  Merck
CAS Number:  18594-05-3
4'-Cyclohexylacetophenone

Glass bottle for solids, 25 g

CAS Number:  18594-05-3
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  p-Cyclohexylacetophenone
  • CAS Number:18594-05-3
  • Formula:C₁₄H₁₈O
  • MDL Number:MFCD00001453
  • Molecular Weight:202.3 g/mol
  • Boiling Pt:316 °C (1013 hPa)
  • Melting Pt:65…67 °C
  • EINECS:242-432-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications