Order Entry
Finland
Orders LinkContactUsLinkComponent
4'-Cyclohexylacetophenone ≥99%

4'-Cyclohexylacetophenone ≥99%

4'-Cyclohexylacetophenone ≥99%
Certificates
4'-Cyclohexylacetophenone ≥99%
4'-Cyclohexylacetophenone ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 18594-05-3 Molecular Formula: C14H18O Formula Weight: 202.30 Melting Point: 66-70° Boiling Point: 179-181°/14mm MDL No.: MFCD00001453

Synonyms:  p-Cyclohexylacetophenone
  • CAS Number:18594-05-3
  • Formula:C₁₄H₁₈O
  • MDL Number:MFCD00001453
  • Molecular Weight:202.3 g/mol
  • Boiling Pt:179…181 °C (14 mmHg)
  • Melting Pt:66…70 °C
  • Storage Temperature:Ambient
  • EINECS:242-432-4
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.