2-Methoxy-4-nitrophenol 97%, Sigma-Aldrich®
Catalog # 326828-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance (Colour):Yellow
- Appearance (Form):Crystal to Powder
- Infrared spectrum:Conforms to Structure
- Carbon:48.0 - 51.4%
- Nitrogen:8.0 - 8.6%
- Purity (GC):≥96.5%
Specification Test Results
About this item
:
:169.14 g/mol
Melting Pt:103…104 °C
EINECS:221-839-0
:GHS07
Storage Temperature:Refrigerator
CAS Number:3251-56-7
Formula:C₇H₇NO₄
MDL Number:MFCD00012143