



Zinc naphthenate (∼8% Zn)
Supplier: TCI
Certificates
About this item
CAS Number:12001-85-3
:
Formula:2(C₁₁H₇O₂)·Zn
MDL Number:MFCD00147699
:319.71 g/mol
Flash Pt:43°C
EINECS:234-409-2
UN:1993
ADR:3,III
:GHS02
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Yellow to Brown clear liquid
- Zinc:7.5 to 8.4 %