p-Xylylenebis(N,N-diethyldithiocarbamate) ≥98.0% (by HPLC, total nitrogen)
TCIAX0056-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:75.0 to 80.0 deg-C
- Solubility in Acetone:almost transparency
:
:400.68 g/mol
:89964-93-2
:C₁₈H₂₈N₂S₄
:MFCD00191696