Order Entry
Finland
ContactUsLinkComponent
3,4,5-Trifluorophenyl trans,trans-4'-Propylbicyclohexyl-4-carboxylate ≥98.0% (by GC)
3,4,5-Trifluorophenyl trans,trans-4'-Propylbicyclohexyl-4-carboxylate ≥98.0% (by GC)
Supplier:  TCI
Certificates
3,4,5-Trifluorophenyl trans,trans-4'-Propylbicyclohexyl-4-carboxylate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Mesomorphic range:
    53 to 120 deg-C

Specification Test Results