
(Trifluoromethyl)cyclohexane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:401-75-2
:
Formula:C₇H₁₁F₃
MDL Number:MFCD03412289
:152.16 g/mol
Boiling Pt:107°C
UN:1993
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %