Trimethylsilyl methacrylate ≥98.0% (by GC) stabilised
: TCI
:13688-56-7
:
:C₇H₁₄O₂Si
:MFCD00053868
:158.27 g/mol
:52°C
:32°C
:1993
:3,III
:Stabilised with N-nitrosodiisopropanolamine
:GHS02
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %