Trihydroxyethylrutin ≥90.0% (by HPLC)
: TCI
:7085-55-4
:
:C₃₃H₄₂O₁₉
:MFCD00893813
:742.7 g/mol
:14,09789
:186°C
:Freezer
:230-389-4
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 90.0 area%
- Specific rotation [a]20/D:-49.0 to -53.0 deg (C=1,Pyridine)